missing translation for 'onlineSavingsMsg'
Learn More

Acycloguanosine, 98%, Thermo Scientific™

Catalog Number p-4813301
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
59277-89-3
C8H11N5O3
225.21
MFCD00057880
MKUXAQIIEYXACX-UHFFFAOYSA-N
acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum
2022
CHEBI:2453
2-amino-9-(2-hydroxyethoxymethyl)-3H-purin-6-one
NC1=NC2=C(N=CN2COCCO)C(=O)N1
This item is not returnable. View return policy
59277-89-3
C8H11N5O3
225.21
MFCD00057880
MKUXAQIIEYXACX-UHFFFAOYSA-N
acyclovir, aciclovir, acycloguanosine, zovirax, vipral, virorax, wellcome-248u, 9-2-hydroxyethoxy methyl guanine, zovir, aciclovirum
2022
CHEBI:2453
2-amino-9-(2-hydroxyethoxymethyl)-3H-purin-6-one
NC1=NC2=C(N=CN2COCCO)C(=O)N1
Melting Point 256°C to 260°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Merck Index 15134
Solubility Information Solubility in water: 0.7mg/mL
Formula Weight 225.2
Percent Purity 98%
Chemical Name or Material Acycloguanosine
compliance-icons
Product Identifier
  • Acycloguanosine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'