Learn More
Socorex™ Acura Variable Volume Manual Pipettes
Secure the stability of your settings when pipetting with Socorex™ Acura Variable Volume Manual Pipettes. Each individual unit's accuracy has been measured for reliability at 0.5μL and 1μL.
Brand:  Socorex™ 825.1000
Code : 04
Additional Details : Weight : 0.18000kg
 View more versions of this product
Product Code. 10794913
  • £267.00 / Each
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
  • Constructed of polypropylene
  • With precision setting mechanism
  • Single-handed volume setting
Specifications
Specifications
Autoclavable
825
Acura™
100 to 1000 μL
Polypropylene
1
Micropipette
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products