Learn More
Socorex™ Acura™ Manual 815 Fixed Volume Micropipettes
Slim and precise micropipettes set at a fixed volume for easy dose repetition. Socorex™ Acura™ Manual 815 Fixed Volume Micropipettes are easy to handle and have a soft plunger stroke.
£169.00
Specifications
Certifications/Compliance | ISO 8655, GMP, GLP, NCCLS, CE IVD Directive 98/79 EEC (in vitro diagnostic medical devices). |
---|---|
Autoclavable | Autoclavable |
Model | Acura™ Manual 815 |
Type | Micropipette |
Quantity | 1 Each |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10263152
|
Socorex™
815.0050 |
50 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10355122
|
Socorex™
815.0100 |
100 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10161582
|
Socorex™
815.0200 |
200 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10724613
|
Socorex™
815.0250 |
250 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10733463
|
Socorex™
815.1000 |
1000 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10754623
|
Socorex™
815.0001 |
1 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10467072
|
Socorex™
815.0040 |
40 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10632673
|
Socorex™
815.0060 |
60 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10375022
|
Socorex™
815.0070 |
70 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10551523
|
Socorex™
815.0075 |
75 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10121442
|
Socorex™
815.0080 |
80 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10345122
|
Socorex™
815.0090 |
90 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10591903
|
Socorex™
815.0120 |
120 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10632863
|
Socorex™
815.0150 |
150 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10375262
|
Socorex™
815.0300 |
300 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10485152
|
Socorex™
815.0400 |
400 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10365312
|
Socorex™
815.0500 |
500 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10089651
|
Socorex™
815.0600 |
600 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10099651
|
Socorex™
815.0700 |
700 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10191722
|
Socorex™
815.0750 |
750 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10315412
|
Socorex™
815.0800 |
800 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10099721
|
Socorex™
815.0900 |
900 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10162062
|
Socorex™
815.0005 |
5 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10611333
|
Socorex™
815.0010 |
10 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10764623
|
Socorex™
815.0015 |
15 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10541333
|
Socorex™
815.0020 |
20 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
10447652
|
Socorex™
815.0025 |
25 μL |
£169.00
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
Description
- Air-displacement fixed-volume pipette.
- Exceptional, user-friendly ergonomics.
- Justip™ adjustable tip ejector fitting most tips.
- Shock, UV-light and autoclaving resistance.
- Ultra-soft plunger stroke.
- Available in a variety of volume models.
Specifications
ISO 8655, GMP, GLP, NCCLS, CE IVD Directive 98/79 EEC (in vitro diagnostic medical devices). | |
Acura™ Manual 815 | |
1 Each |
Autoclavable | |
Micropipette | |
36 Months |