missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Activin RIIB Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Activin RIIB |
---|---|
Clone | 60402 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Activin RIIB Monoclonal specifically detects Activin RIIB in Human samples. It is validated for Western Blot.Specifications
Activin RIIB | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q13705 | |
93 | |
Mouse myeloma cell line NS0-derived recombinant human Activin RIIB Ser19-Thr134 Accession # Q13705 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Activin RIIB in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Activin RIIA is observed. |
60402 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ActivinRIIB, ActR-IIB, ACTRIIB, ACVR2B, EC 2.7.11, EC 2.7.11.30, MGC116908 | |
ACVR2B | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only