Learn More
Invitrogen™ Human Activin A Recombinant Protein
Recombinant Human Activin A is a bioactive protein intended for use in cell culture applications. Activin A is involved in multiple biological processes including inflammation, neural development, and haematopoiesis.
Brand: Invitrogen™ PHC9564
319.20 GBP valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00001kg
Description
Human Activin A Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
- High purity—no interference from other proteins or contaminants
- High biological activity—more results with less protein
- Proven compatibility—Gibco™ proteins bioassayed with Gibco™ media
High purity recombinant bioactive protein
Recombinant Human Activin A is one of our more than 250 available Gibco™ recombinant proteins. To help ensure Gibco™ growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.
High Biological Activity
In-house activity testing of Gibco™ Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Activin A specific activity is determined by the dose dependent proliferation of mouse MPC-11 cells.
Proven Compatibility
Gibco™ proteins are bioassayed with Gibco™ media. Since 1962, Gibco™ has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.
Specifications
Cell Culture | |
Gel-Purified | |
Activin A | |
Human | |
Carrier-Free | |
Inhibition of Proliferation of MPC11 cells | |
Dry Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Neurobiology, Stem Cell Research, Differentiation | |
95 % |
25 kDa | |
10μg | |
116 aa | |
Store in freezer (-5 to -30°C). | |
0.5 ng/ml | |
< 1 EU/ml | |
Mammalian | |
Recombinant, Ligand | |
Activins | |
Gibco™ |