missing translation for 'onlineSavingsMsg'
Learn More

Activated charcoal, NORIT™ SA 2, decolorizing, ACROS Organics™

Brand:  Acros Organics 171530050

Code : 29

Additional Details : CAS Number : 7440-44-0 Weight : 5.05000kg

Quantity 5kg
Packaging Plastic drum

 View more versions of this product

Product Code. 10165773

  • £146.10 / 5kg


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Activated charcoal, NORIT™ SA 2
C
MFCD00133992
7440-44-0
12
Activated Carbon

Specifications

molasses number: < 390
Black
5kg
10% max.
Powder
C
MFCD00133992
Activated Carbon
Activated charcoal, NORIT™ SA 2
3652.0°C
7440-44-0
Plastic drum
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents, soluble matter in hno3: <=3.0%, soluble matter in alcohol: <=0.50%
12
15, 1807
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents, soluble matter in hno3: <=3.0%, soluble matter in alcohol: <=0.50%
Videos
SDS