missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated charcoal, DARCO(R), 4-12 mesh, decolorizing, Thermo Scientific Chemicals
125.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
Specifications
Melting Point | 3652.0°C |
Color | Black |
pH | 3.5 to 6 |
Loss on Drying | 12% max. |
Mesh Size | > 4 mesh: < 5%, < 12 mesh: < 5% |
Packaging | Plastic drum |
Quantity | 3 kg |
Merck Index | 15, 1807 |
Solubility Information | Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5% |
Formula Weight | 12 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'