missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, -100 particle size (mesh), Honeywell Fluka™

Catalog Number p-7120117
Click to view available options
Quantity:
175 g
1 kg
5 kg
Packaging:
Metal Drum
Plastic Bottle
This item is not returnable. View return policy
7440-44-0
C
12.011
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
5462310
CHEBI:27594
carbon
[C]
This item is not returnable. View return policy
7440-44-0
C
12.011
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
5462310
CHEBI:27594
carbon
[C]

powder, -100 particle size (mesh), decolorizing

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 3550°C (literature)
Color Black
Density ca. 350 kg/cu.m
pH 6 to 9
Mesh Size –100 particle size (mesh)
Packaging Plastic Bottle
Quantity 175 g
UN Number NONH for all modes of transport
Formula Weight 12.01g/mol
Physical Form Powder
Chemical Name or Material Activated Charcoal
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'