Learn More
ACT1, clone: 9ACT12, eBioscience™
Rat Monoclonal Antibody
Brand:  Affymetrix eBioscience 14-4040-80
Code : Z2
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15207357
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
For Research Use Only.
Description
Description
The monoclonal antibody 9ACT12 reacts with human Act1 (NF-κB activator 1), also known as CIKS (a connection to IkB kinase and stress-activated kinase). Originally cloned for its ability to activate NF-κB and its direct interaction with IKKγ, Act1 has been subsequently shown to play several roles in immunity. In B cells, it is a negative regulator of CD40 and BAFFR signaling affecting reduced survival. This regulation is through direct interaction of Act1 with CD40 or BAFFR and TRAF3. In addition, Act1 is involved in IL-17R signaling. It interacts with IL-17RA through the homotypic SEFIR interaction domain and is critical for downstream NF-κB- and C/EBP-mediated inflammatory responses.
Specifications
Specifications
ACT1
9ACT12
Unconjugated
Purified
Rat
25μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Nuclear Factor kappa B Activator 1, TRAF3 interacting protein 2, TRAF3IP2, CIKS
IgG1