missing translation for 'onlineSavingsMsg'
Learn More

Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals

Catalog Number p-3735235
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
79-10-7, 150-76-5
C3H4O2
72.06
MFCD00004367
NIXOWILDQLNWCW-UHFFFAOYSA-N
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
6581
CHEBI:18308
prop-2-enoic acid
C=CC(=O)O
79-10-7, 150-76-5
C3H4O2
72.06
MFCD00004367
NIXOWILDQLNWCW-UHFFFAOYSA-N
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
6581
CHEBI:18308
prop-2-enoic acid
C=CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98
Melting Point 13.0°C
Density 1.0500g/mL
Boiling Point 139.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCO2H
Refractive Index 1.4192 to 1.4212
Quantity 250 mL
Beilstein 02, 397
Merck Index 15, 121
Specific Gravity 1.05
Solubility Information Solubility in water: miscible. Other solubilities: miscible with most organic solvents,soluble in acetone
Stabilizer 180 to 220ppm MEHQ
Viscosity 1.3 mPa.s (20°C)
Formula Weight 72.06
Percent Purity 98%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Acrylic acid, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acrylic acid, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'