missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide, 99.9%, Thermo Scientific Chemicals

Catalog Number p-7050337
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
79-06-1
C3H5NO
71.08
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
6579
CHEBI:28619
prop-2-enamide
NC(=O)C=C
This item is not returnable. View return policy
79-06-1
C3H5NO
71.08
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide
6579
CHEBI:28619
prop-2-enamide
NC(=O)C=C

Acrylamide is primarily used in research laboratories for electrophoresis, chromatography, electron microscopy and in the sewage and wastewater treatment industries. It is used in the production of polymers mainly in the pulp and paper industry and textile treatment industry. The largest use for acrylamide is as an intermediate in the production of organic chemicals and in the synthesis of polyacrylamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 82°C to 85°C
Density 1.322
Boiling Point 125°C (25 mm)
Flash Point 138°C (280°F)
Assay Percent Range 99.9%
Quantity 25 g
UN Number UN2074
Beilstein 605349
Sensitivity Air and light sensitive
Merck Index 14,129
Solubility Information Soluble in water,ethanol,ether,and chloroform.
Formula Weight 71.08
Percent Purity 99.9%
Physical Form Crystalline
Chemical Name or Material Acrylamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'