missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrolein diethyl acetal, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Acrolein diethyl acetal is a reagent used for the synthesis of a variety of compounds, including cinnamaldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acrolein diethyl acetal is a reagent used for the synthesis of a variety of compounds, including cinnamaldehydes.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Acrolein diethyl acetal is a reagent used for the synthesis of a variety of compounds, including cinnamaldehydes.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Density | 0.852 |
Boiling Point | 124°C to 125°C |
Flash Point | 13°C (55°F) |
Refractive Index | 1.401 |
Quantity | 10 g |
UN Number | UN2374 |
Beilstein | 1701567 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 130.19 |
Percent Purity | 96% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'