missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pall Acrodisc™ Unit with Mustang S Membrane
Disposable chromatography units with high binding capacities and fast flow rates.
Brand: Pall MSTG25S6
Additional Details : Weight : 0.31350kg
Description
- Mustang S membrane is a strong cation exchanger that effectively binds positively-charged proteins and viral particles
- For research use only
Specifications
Mustang S | |
Non-sterile | |
Blister Pack |
0.8 μm | |
10/Pk. | |
25 mm |