missing translation for 'onlineSavingsMsg'
Learn More

Acriflavine neutral, 13.3-15.8% chlorine, Thermo Scientific Chemicals

Catalog Number 10461061
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
8048-52-0
C27H25ClN6
468.99
MFCD00064307
PEJLNXHANOHNSU-UHFFFAOYSA-N
Acriflavine base, 3, 6-Diamino-10-methylacridinium chloride, Euflavine
3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine chloride
[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
This item is not returnable. View return policy
8048-52-0
C27H25ClN6
468.99
MFCD00064307
PEJLNXHANOHNSU-UHFFFAOYSA-N
Acriflavine base, 3, 6-Diamino-10-methylacridinium chloride, Euflavine
3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine chloride
[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 179°C to 181°C
Color Brown-Red to Orange
Loss on Drying 8% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Beilstein 22, I, 650
Merck Index 15, 115
Solubility Information Solubility in water: 1g/3mL. Other solubilities: incompletely soluble in alcohol,practically insoluble in ether,chloroform and,fixed oils
Formula Weight 469.03
Percent Purity 13.3 to 15.8% (chlorine) (on dried substance)
Physical Form Powder
Chemical Name or Material Acriflavine neutral
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'