missing translation for 'onlineSavingsMsg'
Learn More

Acriflavine hydrochloride, Thermo Scientific Chemicals

Catalog Number 10535501
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
69235-50-3
C27H27Cl3N6
541.91
MFCD00069039
MKLTXAHQKDVBLY-UHFFFAOYSA-N
unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride
53393742
acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride
Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12
69235-50-3
C27H27Cl3N6
541.91
MFCD00069039
MKLTXAHQKDVBLY-UHFFFAOYSA-N
unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride
53393742
acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride
Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12

Acriflavine hydrochloride, C27H28Cl4N6, CAS Number-69235-50-3, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acrifiavine hydrochloride, unii-1s73vw819c, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 260.0°C
Color Brown-Red to Orange-Red
Loss on Drying 7% max. (105°C, 1 hr.)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 g
Beilstein 22,I,65
Merck Index 15,115
Solubility Information Solubility in water: 250-300g/L (20°C). Other solubilities: soluble in alcohol,practically insoluble in ether and chloroform
Physical Form Powder
Chemical Name or Material Acriflavine hydrochloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acriflavine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'