missing translation for 'onlineSavingsMsg'
Learn More

Acridine Orange, Thermo Scientific Chemicals

Catalog Number p-7046114
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
10127-02-3
C17H20Cl3N3Zn
438.10
MFCD00081043,MFCD00081043,MFCD00081043
VADJQOXWNSPOQA-UHFFFAOYSA-L
acridine orange c.i. 46005
71774334
zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
[H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
This item is not returnable. View return policy
10127-02-3
C17H20Cl3N3Zn
438.10
MFCD00081043,MFCD00081043,MFCD00081043
VADJQOXWNSPOQA-UHFFFAOYSA-L
acridine orange c.i. 46005
71774334
zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
[H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C

Fluorescent stain for nucleic acids. An RNA polymerase inhibitorAcridine Orange acts as a lysosomal dye. It is used for cell-cycle studies. It plays an important role as a nucleic acid-selective fluorescent cationic dye, which is useful for cell cycle determination. Further, it serves as an inhibitor of RNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescent stain for nucleic acids. An RNA polymerase inhibitorAcridine Orange acts as a lysosomal dye. It is used for cell-cycle studies. It plays an important role as a nucleic acid-selective fluorescent cationic dye, which is useful for cell cycle determination. Further, it serves as an inhibitor of RNA polymerase.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 5 g
Beilstein 3734978
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 438.1
Chemical Name or Material Acridine Orange

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'