missing translation for 'onlineSavingsMsg'
Learn More

Acridine, 97%, Thermo Scientific Chemicals

Catalog Number p-7043624
Click to view available options
Quantity:
5 g
25 g
260-94-6
C13H9N
179.222
MFCD00005025
DZBUGLKDJFMEHC-UHFFFAOYSA-N
9-azaanthracene, 2,3-benzoquinoline, acrydine, 10-azaanthracene, akridin, dibenzo b,e pyridine, 2,3,5,6-dibenzopyridine, akridin czech, benzo b quinoline
9215
CHEBI:36420
acridine
C1=CC=C2C(=C1)C=C3C=CC=CC3=N2
260-94-6
C13H9N
179.222
MFCD00005025
DZBUGLKDJFMEHC-UHFFFAOYSA-N
9-azaanthracene, 2,3-benzoquinoline, acrydine, 10-azaanthracene, akridin, dibenzo b,e pyridine, 2,3,5,6-dibenzopyridine, akridin czech, benzo b quinoline
9215
CHEBI:36420
acridine
C1=CC=C2C(=C1)C=C3C=CC=CC3=N2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 107°C to 111°C
Density 1.005
Boiling Point 346°C
Quantity 5 g
UN Number UN2713
Beilstein 120200
Merck Index 14,122
Formula Weight 179.22
Percent Purity ≥98%
Chemical Name or Material Acridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'