Learn More
ACPT-II, Tocris Bioscience™
Competitive mGlu receptor antagonist
Brand:  Tocris (Bio-Techne) 1112/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13488553
  • £186.00 / 10mg
Estimated Shipment
01-03-2018
Add to basket
Description
Description
Competitive metabotropic receptor antagonist (KB values are 115, 88 and 77 μM at mGlu1a, mGlu2 and mGlu4a respectively).
Specifications
Chemical Identifiers
ACPT-II
C8H11NO6
217.18
(1R,3R,4S)-1-Aminocyclopentane-1,3,4-tricarboxylic acid
Specifications
ACPT-II
10mg
C8H11NO6
(1R,3R,4S)-1-Aminocyclopentane-1,3,4-tricarboxylic acid
195209-04-2
>98%
217.18
Videos