missing translation for 'onlineSavingsMsg'
Learn More

ACPT-I, Tocris Bioscience™

Group III mGlu agonist

Brand:  Tocris (Bio-Techne) 1111/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13458553

  • £149.85 / 10mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Agonist for group III mGlu receptors (EC50 values are 7.2 and 8.2 μM for mGlu4a and mGlu8 respectively). Anticonvulsant in mice.
Specifications

Chemical Identifiers

194918-76-8
217.177
acpt-i, acpt-ii, 1-aminocyclopentane-1,3,4-tricarboxylic acid, 1r,2s-4-aminocyclopentane-1,2,4-tricarboxylic acid, 4-amino-cyclopentane-1,2,4-tricarboxylic acid, tocris-1113, d0w3py, 1s,3r,4s-1-aminocyclopentane-1,3,4-tricarboxylic acid, 1r,2s,4s-4-aminocyclopentane-1,2,4-tricarboxylic acid, 1r,3r,4s-1-aminocyclopentane-1,3,4-tricarboxylic acid
(1R,2S)-4-aminocyclopentane-1,2,4-tricarboxylic acid
C8H11NO6
FERIKTBTNCSGJS-OBLUMXEWSA-N
6604820
C1C(C(CC1(C(=O)O)N)C(=O)O)C(=O)O

Specifications

ACPT-I
194918-76-8
acpt-i, acpt-ii, 1-aminocyclopentane-1,3,4-tricarboxylic acid, 1r,2s-4-aminocyclopentane-1,2,4-tricarboxylic acid, 4-amino-cyclopentane-1,2,4-tricarboxylic acid, tocris-1113, d0w3py, 1s,3r,4s-1-aminocyclopentane-1,3,4-tricarboxylic acid, 1r,2s,4s-4-aminocyclopentane-1,2,4-tricarboxylic acid, 1r,3r,4s-1-aminocyclopentane-1,3,4-tricarboxylic acid
C1C(C(CC1(C(=O)O)N)C(=O)O)C(=O)O
217.177
217.18
10mg
C8H11NO6
FERIKTBTNCSGJS-OBLUMXEWSA-N
(1R,2S)-4-aminocyclopentane-1,2,4-tricarboxylic acid
6604820
>98%
Videos