missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ ACOX1 Recombinant Human Protein, N-His tag

Expressed from DNA sequence encoding the human ACOX1 (AAH08767.1) (Met 1-Leu 660) fused to a polyhistidine tag at the N-terminus

Brand:  Sino Biological™ 11266H07B50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218475

  • £705.80 / 50µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

  • Formulation: 20mM Tris, 500mM NaCl, 20% Glycerol, 3mM DTT, pH 7.0
Specifications

Specifications

Lyophilized
76.7kd
50μg
Full Length, Recombinant
Peroxisomal Acyl-Coenzyme A Oxidase 1
Assay Standard
Insect
His Tag
51
≥90% by SDS-PAGE
Human
<1 EU/μg
ACOX1
Other proteins
ACOX1 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.