Acetyl chloride, ACS reagent, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 447171000

UNSPSC: 12161500

Code: A0

Additional Details:
Additional Details: CAS Number: 75-36-5 Weight: 0.11000kg Transport: UN number : 1717 Chem class : 1a Pack group : II


Packaging Glass bottle
Quantity 100mL

Product Code. 12625067

Quantity Price
1 £ 15.4 / 100mL
EU Stock 8
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Acetyl chloride
CAS 75-36-5
Purity Grade ACS Reagent Grade
Molecular Formula C2H3ClO
Linear Formula CH3COCl
Formula Weight 78.5

Specification

Chemical Name or Material Acetyl chloride
CAS 75-36-5
Health Hazard 1 Danger
Packaging Glass bottle
Purity Grade ACS Reagent Grade
Molecular Formula C2H3ClO
Linear Formula CH3COCl
Formula Weight 78.5
Density 1.1000g/mL
Quantity 100mL
Documentation

Technical documents

Safety and Handling

Danger