missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl bromide, 98%, Thermo Scientific Chemicals
92.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Acetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.
Solubility
Miscible with ether, chloroform, benzene and acetone.
Notes
Incompatible with strong oxidizing agents, strong bases and alcohols
Acetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.
Solubility
Miscible with ether, chloroform, benzene and acetone.
Notes
Incompatible with strong oxidizing agents, strong bases and alcohols
Specifications
Melting Point | -96°C |
Density | 1.66 |
Boiling Point | 75°C to 77°C |
Flash Point | >75°C (167°F) |
Assay Percent Range | ≥98% |
Odor | Acrid |
Linear Formula | CH3COBr |
Refractive Index | 1.45 |
Quantity | 500 g |
UN Number | UN1716 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'