missing translation for 'onlineSavingsMsg'
Learn More

Acetonylacetone, 97%, Thermo Scientific Chemicals

Catalog Number 10578090
75.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
110-13-4
C6H10O2
114.14
MFCD00008792
OJVAMHKKJGICOG-UHFFFAOYSA-N
2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
8035
CHEBI:85014
hexane-2,5-dione
CC(=O)CCC(=O)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
110-13-4
C6H10O2
114.14
MFCD00008792
OJVAMHKKJGICOG-UHFFFAOYSA-N
2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
8035
CHEBI:85014
hexane-2,5-dione
CC(=O)CCC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -6.0°C
Color Yellow to Brown
Density 0.9730g/mL
Boiling Point 185.0°C to 193.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3COCH2CH2COCH3
Refractive Index 1.4250 to 1.4270
Quantity 100 mL
Beilstein 01,788
Merck Index 15,7
Specific Gravity 0.973
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, ether, ethanol and, diethyl ether, miscible in organic solvents
Viscosity 1.43 mPa.s (25°C)
Formula Weight 114.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Acetonylacetone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonylacetone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'