missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, HPLC for Electrochemical Detection, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Melting Point | -46°C |
Color | Colorless |
Boiling Point | 81°C |
Quantity | 2.5 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 97mbar at 20°C |
Viscosity | 0.36cp at 20°C |
Formula Weight | 41.05g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'