Learn More
Acetonitrile, for HPLC, Fisher Chemical™
C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, CHEBI:38472, Colorless, 41.05g/mol, WEVYAHXRMPXWCK-UHFFFAOYSA-N, acetonitrile, -46 deg.C, 1878, 41.053, Amber glass bottle, Liquid, 6342, CC#N, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C
Brand: Fisher Chemical A/0626/PB17
141.96 GBP valid until 2022-06-30
Use promo code "16496" to get your promotional price.
Code : 25
Additional Details : CAS Number : 75-05-8 Weight : 1.96500kg Transport : UN number : 1648 Chem class : 3 Pack group : II
Packaging | Plastic Coated Glass bottle |
---|---|
Quantity | 2.5L |
Description
C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, CHEBI:38472, Colorless, 41.05g/mol, WEVYAHXRMPXWCK-UHFFFAOYSA-N, acetonitrile, -46 deg.C, 1878, 41.053, Amber glass bottle, Liquid, 6342, CC#N, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C
Chemical Identifiers
75-05-8 | |
C2H3N | |
1878 |
Specifications
75-05-8 | |
1878 | |
Liquid | |
Colorless | |
2.5L | |
1.42 | |
0.36cp at 20°C |
C2H3N | |
41.05g/mol | |
81°C | |
-46°C | |
Plastic Coated Glass bottle | |
97mbar at 20°C |
Safety and Handling
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.