missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, far UV/gradient grade, 99.9+%, Thermo Scientific Chemicals
£95.10 - £198.00
Specifications
Melting Point | −48°C to −45°C |
---|---|
CAS | 75-05-8 |
Boiling Point | 80°C to 82°C |
Molecular Formula | C{2}H{3}N |
Synonym | Methyl cyanide, Cyanomethane |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15416207
|
GC Internal (Alfa Aesar)
H33387.AU |
1000 mL |
£95.10
1000 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15406207
|
Thermo Scientific Alfa Aesar
H33387.0F |
2500 mL |
£198.00
2500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
−48°C to −45°C | |
80°C to 82°C | |
Methyl cyanide, Cyanomethane |
75-05-8 | |
C{2}H{3}N | |
99.9+% |