Learn More
Acetonitrile, for DNA Synthesis and Protein Sequencing, Fisher Chemical™
C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Amber glass bottle, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C
Brand: Fisher Chemical A/0628/15
Code : 22
Additional Details : CAS Number : 75-05-8 Weight : 0.78600kg Transport : UN number : 1648 Chem class : 3 Pack group : II
Description
C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Amber glass bottle, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C
Specifications
75-05-8 | |
1878 | |
Liquid | |
97mbar at 20°C | |
0.36cp at 20°C | |
Colorless | |
1L |
C2H3N | |
41.05g/mol | |
Amber glass bottle | |
1.42 | |
81°C | |
-46°C |
Safety and Handling
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.