missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile-d3, 99.8% (Isotopic), Thermo Scientific Chemicals

Catalog Number p-7051924
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2206-26-0
C2H3N
44.07
MFCD00001881
WEVYAHXRMPXWCK-FIBGUPNXSA-N
acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
123151
2,2,2-trideuterioacetonitrile
[2H]C([2H])([2H])C#N
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2206-26-0
C2H3N
44.07
MFCD00001881
WEVYAHXRMPXWCK-FIBGUPNXSA-N
acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
123151
2,2,2-trideuterioacetonitrile
[2H]C([2H])([2H])C#N

Acetonitrile d3 is used as a solvent for NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 0.844g/mL
Flash Point 5° (42°F)
Odor Pungent
Sensitivity Hygroscopic
Quantity 2 g
Boiling Point 81°C
UN Number UN1648
Merck Index 14,70
Formula Weight 44.08
Percent Purity 99.8%
Chemical Name or Material Acetonitrile-d3, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'