missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Certified AR for Analysis, Fisher Chemical™

Catalog Number 10345070
280.92 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Metal Can
Plastic-coated Glass Bottle
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
75-05-8
C2H3N
41.05
1878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
CC#N
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
75-05-8
C2H3N
41.05
1878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
CC#N

C2H3N, CAS Number-75-05-8, 5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Metal can, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C

TRUSTED_SUSTAINABILITY
Melting Point -46°C
Color Colorless
Boiling Point 81°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 97mbar at 20°C
Viscosity 0.36cp at 20°C
Formula Weight 41.05g/mol
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'