missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Acetone semicarbazone, 96%

Brand:  Alfa Aesar™ A12436.18

Code : NEW

Additional Details : CAS Number : 110-20-3 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 11354619

  • £13.60 / 50g


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Specifications

Chemical Identifiers

Acetone semicarbazone
C4H9N3O
MFCD00014785
14,8444
110-20-3
115.14
970110

Specifications

Acetone semicarbazone
96%
115.14
970110
179° to 185°C
110-20-3
C4H9N3O
MFCD00014785
14,8444
50g
Videos
SDS