missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Extra Dry, for Synthesis, Fisher Chemical™

Brand:  Fisher Chemical A/0602/17

Code : 25

Additional Details : CAS Number : 67-64-1 Weight : 1.97750kg Transport : UN number : 1090 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 10151510

  • £122.00 / 2.50 L

In stock, ready to ship: 124

Please to purchase this item. Need a web account? Register with us today!


For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Description

Description

Specifications

Specifications

67-64-1
8765
Liquid
247mbar at 20°C
2
-95°C
Colorless
C3H6O
58.08g/mol
Amber glass bottle
0.32 mPaS at 20°C
7
56°C
2.5L
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetone

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates
View accessory products