missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Acetone-d6, 99.9% (Isotopic), contains 1% v/v TMS

Brand:  Alfa Aesar™ 042458.14

Code : NEW

Additional Details : CAS Number : 666-52-4 Weight : 0.02500kg Transport : UN number : 1090 Chem class : 3 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 11314618

  • £68.80 / 25g


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Specifications

Chemical Identifiers

Acetone-d6
Isotopic
MFCD00044635
666-52-4
64.13
UN1090

Specifications

Acetone-d6
0.872g/mL
Acetone-like
56°C
−94°C
Isotopic
Liquid
64.13
UN1090
Contains 1% v/v TMS
−17°C (1°F)
1.3554
Colorless
25g
666-52-4
99.9%
MFCD00044635
Videos
SDS