missing translation for 'onlineSavingsMsg'
Learn More

Acetone, For pesticide residue analysis, Honeywell Riedel-de Haën™

Catalog Number 15644190
184.90 GBP valid until 2024-12-31
Use promo code "21898" to get your promotional price.


184.90 GBP valid until 2024-12-31
Use promo code "21905" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
180
CHEBI:15347
propan-2-one
CC(C)=O
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
180
CHEBI:15347
propan-2-one
CC(C)=O

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY
Melting Point -94°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 0.791 g/mL at 25 °C(lit.)
Boiling Point 55°C to 57°C
Quantity 4 x 2.5 L
Flash Point −17°C
Packaging Glass Bottle
Linear Formula CH3COCH3
UN Number UN1090
Beilstein 635680
Formula Weight 58.08
Percent Purity >99.8%
Grade Pesticide
Chemical Name or Material Acetone, For pesticide residue analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'