missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetanilide, 98+%, Thermo Scientific Chemicals

Catalog Number p-7023713
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
102-01-2
C10H11NO2
177.203
MFCD00008780
DYRDKSSFIWVSNM-UHFFFAOYSA-N
acetoacetanilide, butanamide, 3-oxo-n-phenyl, acetoacetic anilide, acetoacetamidobenzene, acetylacetanilide, acetoacetylaniline, acetoacetanilid, n-phenylacetoacetamide, acetoacetic acid anilide, n-acetylacetyl aniline
7592
3-oxo-N-phenylbutanamide
CC(=O)CC(=O)NC1=CC=CC=C1
This item is not returnable. View return policy
102-01-2
C10H11NO2
177.203
MFCD00008780
DYRDKSSFIWVSNM-UHFFFAOYSA-N
acetoacetanilide, butanamide, 3-oxo-n-phenyl, acetoacetic anilide, acetoacetamidobenzene, acetylacetanilide, acetoacetylaniline, acetoacetanilid, n-phenylacetoacetamide, acetoacetic acid anilide, n-acetylacetyl aniline
7592
3-oxo-N-phenylbutanamide
CC(=O)CC(=O)NC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 83°C to 86°C
Density 1.26
Boiling Point 129°C (24 mmHg)
Flash Point 150°C (302°F)
Odor Odorless
Quantity 250 g
Beilstein 473419
Merck Index 14,58
Formula Weight 177.2
Percent Purity ≥98%
Chemical Name or Material Acetoacetanilide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'