missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, TraceSELECT™, ≥99%, Honeywell Fluka™

Catalog Number p-7120100
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Poly Bottle
This item is not returnable. View return policy
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O
This item is not returnable. View return policy
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O

TraceSELECT™, for trace analysis,≥99.0%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY
Melting Point 16.2°C
Boiling Point 117°C to 118°C
Color Colorless
Appearance Liquid
Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05g/mol
Percent Purity ≥99%
Grade TraceSELECT™
Density 1.049g/mL at 25°C
Quantity 100 mL
Explosion Limit 4% at 59°F, 16% at 92°F
Flash Point 40°C (104°F)
Ignition Point 800°F
Packaging Poly Bottle
Refractive Index n20/D 1.371
Vapor Density 2.07 (vs air)
Chemical Name or Material Acetic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'