missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt trihydrate, 99+%, for biochemistry, ACROS Organics™

Brand:  Acros Organics 446790050

Code : 29

Additional Details : CAS Number : 6131-90-4 Weight : 5.05000kg

Quantity 5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 12193751

  • £172.60 / 5kg


Estimated Shipment
16-05-2018
Add to basket

Specifications

Chemical Identifiers

Acetic acid, sodium salt trihydrate
For biochemistry
C2H3NaO2·3H2O
136.08
6131-90-4
99+%
CH3COONa·3H2O
Sodium acetate trihydrate

Specifications

0.05 max. (3M) at 260nm, 0.05 max. (3M) at 280nm
6131-90-4
99+%
C2H3NaO2·3H2O
136.08
Sodium acetate trihydrate
5kg
Acetic acid, sodium salt trihydrate
For biochemistry
99+%
CH3COONa·3H2O
Plastic bottle
58.0°C
Videos
SDS