missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, sodium salt, 99+%, for mol biology, anhydrous, DNAse, RNAse and P, Thermo Scientific Chemicals
27.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Absorbance | (0.1 M in water), (0.1 M in water) at 0nm, 0.01 max. at 260nm, 0.01 max. at 280nm |
pH | 7 to 9.2 (5% soln. at 25°C) |
Melting Point | 324.0°C |
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Color | White |
Assay Percent Range | 99+% |
Linear Formula | CH3CO2Na |
Solubility Information | Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol |
IUPAC Name | sodium;acetate |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'