missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, ACROS Organics™

Brand:  Acros Organics 424250050

Code : 29

Additional Details : CAS Number : 127-09-3 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10607601

  • £13.00 / 5g


Estimated Shipment
17-05-2018
Add to basket

Specifications

Chemical Identifiers

Acetic acid, sodium salt
99.0
ACS Reagent Grade
99+%
CH3CO2Na
MFCD00012459
127-09-3
100.0
≥99%
C2H3NaO2
82.03
Sodium acetate

Specifications

Acetic acid, sodium salt
99.0
ACS Reagent Grade
Fine Crystalline Powder
99+%
C2H3NaO2
82.03
02, 96
15, 8709
10ppm max.
0.01% max.
1.0% max. (120°C)
Glass bottle
Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
324.0°C
White
>250°C
127-09-3
100.0
50ppm max.
≥99%
20ppm max.
CH3CO2Na
MFCD00012459
01,1024; 05,591
Sodium acetate
Authentic
10ppm max.
20ppm max.
10ppm max.
30ppm max.
7.0 to 9.2 (5% soln. at 25°C)
5g
Videos
SDS