missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™

Catalog Number 15643920
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
60 kg
6 x 1 L
200 L
Packaging:
Glass Bottle
Plastic Bottle
Plastic Pail
Poly Drum
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05g/mol
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Density 1.049g/mL at 25°C
Quantity 1 L
Explosion Limit 16% at 92°F, 4% at 59°F
Flash Point 40°C (104°F)
Ignition Point 800°F
Packaging Glass Bottle
Vapor Density 2.07 (vs air)
Chemical Name or Material Acetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'