missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™
227.56 GBP valid until 2024-03-31
Use promo code "21663" to get your promotional price.
Use promo code "21663" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
60 kg
6 x 1 L
200 L
Packaging:
Glass Bottle
Plastic Bottle
Plastic Pail
Poly Drum
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Linear Formula | CH3CO2H |
UN Number | UN2789 |
Formula Weight | 60.05 |
Percent Purity | ≥99.8% |
Density | 1.049g/mL at 25°C |
Quantity | 4 x 2.5 L |
Explosion Limit | 16% at 92°F, 4% at 59°F |
Flash Point | 40°C (104°F) |
Ignition Point | 800°F |
Packaging | Glass Bottle |
Show More |
Safety and Handling
Product Identifier
- Acetic acid
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'