missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.8-100.5%, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 kg
30 kg
4 x 2.5 L
60 kg
6 x 1 L
200 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 16.2°C (literature) |
Boiling Point | 118°C |
CAS Max % | 100.0000% |
Assay Percent Range | 99.8 to 100.5% |
Linear Formula | CH3CO2H |
UN Number | UN2789 |
Beilstein | 506007 |
Formula Weight | 60.05 |
Percent Purity | 99.8 to 100.5% |
Grade | puriss. |
Show More |
Safety and Handling
Product Identifier
- Acetic acid
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'