missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, potassium salt, 99+%, for biochemistry, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Melting Point | 292.0°C |
Flash Point | >250°C |
Assay Percent Range | 99+% |
Packaging | Plastic bottle |
Linear Formula | CH3CO2K |
Quantity | 500 g |
Merck Index | 15,7726 |
Solubility Information | (5 M in water) Clear colorless solution |
Formula Weight | 98.14 |
Percent Purity | 99+% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'