missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, potassium salt, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-4509635
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
127-08-2
C2H3KO2
98.142
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]
127-08-2
C2H3KO2
98.142
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
517044
CHEBI:32029
potassium;acetate
CC(=O)[O-].[K+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
pH 6.5 to 9.0 (5% soln. at 25°C)
Melting Point 292.0°C
Color White
Assay Percent Range 99+%
Linear Formula CH3CO2K
Beilstein 02, 96
Fieser 01,906; 06,475
Merck Index 15, 7726
Solubility Information Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 98.14
Percent Purity ≥99%
Grade ACS Reagent
Quantity 500 g
Flash Point >250°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Chemical Name or Material Acetic acid, potassium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'