missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial USP/FCC/ACS, Macron Fine Chemicals™
92.06 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
Packaging:
SAFEMOR
Specifications
Formula Weight | 60.05 |
Grade | ACS/FCC/USP |
Quantity | 500 mL |
Packaging | SAFEMOR |
Chemical Name or Material | Acetic Acid, Glacial |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'