missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial, BAKER INSTRA-ANALYZED Plus, J.T. Baker™
324.90 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Description
BAKER INSTRA-ANALYZED™ Plus acids allow for testing of more trace metals with tighter specifications.
- Quality assured for up to 64 trace metals tested to low parts-per-billion (ppb) levels
- Lot to lot consistency ensures reliable results between tests
- Space-saving HDPE packaging with low metallic extractables
- Enhanced safety with tamper-evident caps
- Environmentally-friendly recyclable HDPE bottles
- Strict protocols in an ISO facility ensure quality in manufacturing
Specifications
pH | 2.4 (1.0M aq. soln.) |
Melting Point | 16.4°C |
Boiling Point | 117.9°C |
Color | Colorless |
Linear Formula | CH3CO2H |
Solubility Information | Miscible in water |
Formula Weight | 60.05g/mol |
Density | 1.05g/mL at 20°C |
Percent Purity | 99% (min.) |
Grade | For use in Trace Metal Analysis |
Show More |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'