missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, glacial, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
20 L
4 x 4 L
Description
Good polar protic solvent, in its ester form it acts as a solvent for inks paints and coatings. Used in laboratory as a solvent for recrystallization, manufacture of camphor, cellulose acetate, polyvinyl acetate. It involves in Wagner-Meerwein rearrangement. Used for preparation of mono-lithium salt of 5-sulfoisophthalic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
pH | 2.5 |
Melting Point | 16.6°C |
Boiling Point | 118.1°C |
UN Number | UN2789 |
Merck Index | 14,55 |
Solubility Information | Miscible with water,ethanol,acetone,toluene and immiscible with carbon disulphide. |
Formula Weight | 60.05 |
Percent Purity | ≥99% |
Density | 1.049 |
Quantity | 20 L |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'