missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, ammonium salt, 98+%, for analysis, ACROS Organics™

Brand:  Acros Organics 218365000

Code : 29

Additional Details : CAS Number : 631-61-8 Weight : 0.55000kg

Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10543961

  • £33.50 / 500g


EU Stock 29
Add to basket

Description

Description

Biological buffer for analysis
Specifications

Chemical Identifiers

Acetic acid, ammonium salt
631-61-8
100.0
≥98%
C2H7NO2
77.08
Anhydrous
98.0
For analysis
98+%
CH3CO2NH4
MFCD00013066

Specifications

Acetic acid, ammonium salt
631-61-8
100.0
For analysis
≥98%
20ppm max.
CH3CO2NH4
MFCD00013066
01,38; 07,11
Ammonium acetate
Authentic
20ppm max.
Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C)
3% Max.
6.5 to 7.5 (5% soln. at 20°C)
500g
Anhydrous
98.0
0.01% max.
Crystals or Crystalline Powder
98+%
C2H7NO2
77.08
02, 96
15, 492
20ppm max.
20ppm max.
Plastic bottle
20ppm max.
114.0°C
White
136°C
Videos
SDS