missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, 1.0N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
20 L
Description
Acetic acid, 1.0N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 20 L |
Formula Weight | 60.05 |
Solubility | Miscible with water |
Physical Form | Liquid |
Chemical Name or Material | Acetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'