missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, 1.0N Standardized Solution, Alfa Aesar™

Brand:  Alfa Aesar 035566.K7

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 4L

 View more versions of this product

Product Code. 11303667

  • £50.90 / 4 L


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Acetic acid
CH3CO2H
MFCD00036152
Liquid
60.05

Specifications

Acetic acid
Liquid
60.05
4L
Liquid
CH3CO2H
MFCD00036152
Videos
SDS