missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetazolamide, 99%, Thermo Scientific Chemicals
14.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Acetazolamide acts as a carbonic anhydrase inhibitor which increases cerebral blood flow. It inhibits water permeability of membranes by interacting with aquaporins. It is used for the medical treatment of glaucoma, epileptic seizure, idiopathic intracranial hypertension, altitude sickness, cystinuria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in NH4OH (50 mg/mL), DMSO, Methanol and slightly soluble in Ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
Soluble in NH4OH (50 mg/mL), DMSO, Methanol and slightly soluble in Ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
Specifications
Melting Point | ∽255°C (decomposition) |
Quantity | 5 g |
Beilstein | 212994 |
Merck Index | 14,53 |
Solubility Information | Soluble in NH4OH (50mg/mL),DMSO,Methanol and slightly soluble in Ethanol. |
Formula Weight | 222.25 |
Percent Purity | 99% |
Chemical Name or Material | Acetazolamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'